missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CCR2 Partial ORF (NP_000639, 1 a.a. - 42 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00001231-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
Sequence: MLSTSRSRFIRNTNESGEEVTTFFDYDYGAPCHKFDVKQIGASpécification
NP_000639 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
30.36kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLSTSRSRFIRNTNESGEEVTTFFDYDYGAPCHKFDVKQIGA | |
RUO | |
CCR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1231 | |
CCR2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CC-CKR-2/CCR2A/CCR2B/CD192/CKR2/CKR2A/CKR2B/CMKBR2/MCP-1-R | |
CCR2 | |
Recombinant | |
wheat germ expression system |