Learn More
ACROBiosystems Human CD161 Protein, Fc Tag
sur devis
Spécification
Poids moléculaire | 44.9 kDa |
---|---|
Source | Human CD161, Fc Tag (CD1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 67 - Ser 225 (Accession # Q12918-1). |
Immunogène | Gln 67 - Ser 225 |
Conditions de stockage | -20°C |
Alias de gène | CD161, C-type lectin domain family 5 member B, HNKR-P1a, NKR-P1A, Natural killer cell surface protein P1A |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17246080
|
ACROBiosystems
CD1-H5253-1MG |
1 mg |
sur devis
1mg |
N/A | Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Cet article nécessite une mise à jour du prix avant d'être commandé. Demandez un prix pour cet article ici. |
|||||||||
17236080
|
ACROBiosystems
CD1-H5253-100UG |
100 μg |
sur devis
100µg |
N/A | Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Cet article nécessite une mise à jour du prix avant d'être commandé. Demandez un prix pour cet article ici. |
|||||||||
Description
CD161 is also known as NKR-P1A, CLEC5B and KLRB1which plays an inhibitory role on natural killer (NK) cells cytotoxicity. Activation results in specific acid sphingomyelinase/SMPD1 stimulation with subsequent marked elevation of intracellular ceramide. Activation also leads to AKT1/PKB and RPS6KA1/RSK1 kinases stimulation as well as markedly enhanced T-cell proliferation induced by anti-CD3. Acts as a lectin that binds to the terminal carbohydrate Gal-alpha Gal epitope as well as to the N-acetyllactosamine epitope. Binds also to CLEC2D/LLT1 as a ligand and inhibits NK cell-mediated cytotoxicity as well as interferon-gamma secretion in target cells. New research shows that genetic inactivation of KLRB1 or antibody-mediated CD161 blockade enhances T cell-mediated killing of glioma cells in vitro and their anti-tumor function in vivo. KLRB1 and its associated transcriptional program are also expressed by substantial T cell populations in other human cancers. The reference provides an atlas of T cells in gliomas and highlights CD161 and other NK cell receptors as immunotherapy targets.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 44.9 kDa.Spécification
44.9 kDa | |
Gln 67 - Ser 225 | |
CD161, C-type lectin domain family 5 member B, HNKR-P1a, NKR-P1A, Natural killer cell surface protein P1A | |
Unconjugated | |
Fc Tag | |
Powder |
Human CD161, Fc Tag (CD1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 67 - Ser 225 (Accession # Q12918-1). | |
-20°C | |
CD161 | |
Human | |
HEK293 | |
Immune Checkpoint Proteins |