missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human CD28H / IGPR-1 Protein, Fc Tag
423.38€ - 2818.49€
Spécification
Numéro d’adhésion | AAH15655.1 |
---|---|
Poids moléculaire | 40.6 kDa |
Source | Human CD28H, Fc Tag (CDH-H5251) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 150 (Accession # AAH15655.1). |
Immunogène | Leu 23 - Gly 150 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15846407
|
ACROBiosystems
CDH-H5251-1MG |
1 mg |
2818.49€
1mg |
Expédition estimée: 27-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15836407
|
ACROBiosystems
CDH-H5251-100UG |
100 μg |
423.38€
100µg |
Expédition estimée: 27-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
CD28 homolog (CD28H), also called transmembrane and immunoglobulin domain containing 2 (TMIGD2) and IGPR-1,is encoded by the TMIGD2 gene. CD28H is constitutively expressed on naive T and NK cells. After interaction of CD28 homolog with B7 homologue,the peripheral effector and memory T cells can be activated and proliferated by Akt-dependent signalling cascade. Plays a role in cell-cell interaction, cell migration, and angiogenesis. Through interaction with HHLA2, costimulates T-cells in the context of TCR-mediated activation. Enhances T-cell proliferation and cytokine production via an AKT-dependent signaling cascade.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.6 kDa. As a result of glycosylation, the protein migrates as 45-60 kDa under reducing (R) condition, and 90-120 kDa under non-reducing (NR) condition (SDS-PASpécification
AAH15655.1 | |
Human CD28H, Fc Tag (CDH-H5251) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 150 (Accession # AAH15655.1). | |
-20°C | |
CD28H | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
40.6 kDa | |
Leu 23 - Gly 150 | |
TMIGD2,IGPR1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |