missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CHL1 Partial ORF (NP_006605, 26 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00010752-Q01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16153116

  • 545.00€ / 25µg
Expédition estimée: 04-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a member of the L1 gene family of neural cell adhesion molecules. It is a neural recognition molecule that may be involved in signal transduction pathways. The deletion of one copy of this gene may be responsible for mental defects in patients with 3p- syndrome. Several alternatively spliced transcript variants of this gene have been described, but their full length nature is not known. [provided by RefSeq]

Sequence: EIPSSVQQVPTIIKQSKVQVAFPFDEYFQIECEAKGNPEPTFSWTKDGNPFYFTDHRIIPSNNSGTFRIPNEGHISHFQGKYRCFASNKLGIAMSEEIEFIVPSVPKFPK
Spécification

Spécification

NP_006605
Liquid
10752
CHL1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CALL/FLJ44930/L1CAM2/MGC132578
CHL1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EIPSSVQQVPTIIKQSKVQVAFPFDEYFQIECEAKGNPEPTFSWTKDGNPFYFTDHRIIPSNNSGTFRIPNEGHISHFQGKYRCFASNKLGIAMSEEIEFIVPSVPKFPK
RUO
CHL1
Wheat Germ (in vitro)
GST
Vidéos
FDS
Documents
Certificats
Promotions

Promotions