missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CSNK2A2 Partial ORF (NP_001887.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00001459-Q01.10ug
Informations supplémentaires : Poids : 0.02000kg
Description
Sequence: MPGPAAGSRARVYAEVNSLRSREYWDYEAHVPSWGNQDDYQLVRKLGRGKYSEVFEAINITNNERVVVKILKPVKKKKIKREVKILENLRGGTNIIKLIDSpécification
NP_001887.1 | |
Liquid | |
1459 | |
CSNK2A2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CK2A2/CSNK2A1/FLJ43934 | |
CSNK2A2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPGPAAGSRARVYAEVNSLRSREYWDYEAHVPSWGNQDDYQLVRKLGRGKYSEVFEAINITNNERVVVKILKPVKKKKIKREVKILENLRGGTNIIKLID | |
RUO | |
CSNK2A2 | |
Wheat Germ (in vitro) | |
GST |