Learn More
Abnova™ Human DDX3Y Partial ORF (NP_004651, 1 a.a. - 80 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, and it has a homolog on the X chromosome. The gene mutation causes male infertility, Sertoli cell-only syndrome or severe hypospermatogenesis, suggesting that this gene plays a key role in the spermatogenic process. Alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]
Spécification
Spécification
Numéro d’adhésion | NP_004651 |
À utiliser avec (application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Identification génétique (Entrez) | 8653 |
Poids moléculaire | 34.54kDa |
Nom | DDX3Y (Human) Recombinant Protein (Q01) |
Test du contrôle qualité | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantité | 10 μg |
Immunogène | MSHVVVKNDPELDQQLANLDLNSEKQSGGASTASKGRYIPPHLRNREASKGFHDKDSSGWSCSKDKDAYSSFGSRDSRGK |
Conditions de stockage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Afficher plus |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.