Learn More
Abnova™ Human EFS Partial ORF (AAH34246, 369 a.a. - 468 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00010278-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene contains a SH3 domain, which is known to be important in intracellular signal transduction. The protein encoded by a similiar gene in mice was shown to be able to bind to SH3 domain of protein-tyrosine kinases. The function of this gene is unknown. Two alternatively spliced variants have been described. [provided by RefSeq]
Sequence: QANQPPRLFVPHSKRVVVAAHRLVFVGDTLGRLAASAPLRAQVRAAGTALGQALRATVLAVKGAALGYPSSPAIQEMVQCVTELAGQALQFTTLLTSLAPSpécification
AAH34246 | |
Liquid | |
10278 | |
EFS (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAS3/CASS3/EFS1/EFS2/HEFS/SIN | |
EFS | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QANQPPRLFVPHSKRVVVAAHRLVFVGDTLGRLAASAPLRAQVRAAGTALGQALRATVLAVKGAALGYPSSPAIQEMVQCVTELAGQALQFTTLLTSLAP | |
RUO | |
EFS | |
Wheat Germ (in vitro) | |
GST |