missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Ephrin-A4 / EFNA4 Protein, His Tag
314.52€ - 2153.18€
Spécification
Numéro d’adhésion | AAI07484 |
---|---|
Poids moléculaire | 17.5 kDa |
Source | Human Ephrin-A4, His Tag (EA4-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gly 171 (Accession # AAI07484). |
Immunogène | Leu 26 - Gly 171 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15876597
|
ACROBiosystems
EA4-H5229-100UG |
100 μg |
314.52€
100µg |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16467479
|
ACROBiosystems
EA4-H5229-1MG |
1 mg |
2153.18€
1mg |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Ephrin-A4 (EFNA4) is also known as EPH-related receptor tyrosine kinase ligand 4 (LERK4), EPLG4, which belongs to the ephrin family. EFNA4 contains one ephrin RBD (ephrin receptor-binding) domain. EFNA4 is cell surface GPI-bound ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. EFNA4 binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. EFNA4 may play a role in the interaction between activated B-lymphocytes and dendritic cells in tonsils.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.5 kDa. The protein migrates as 18-20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAI07484 | |
Human Ephrin-A4, His Tag (EA4-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gly 171 (Accession # AAI07484). | |
-20°C | |
Ephrin-A4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
17.5 kDa | |
Leu 26 - Gly 171 | |
EFNA4,EPLG4,LERK4,Ephrin-A4 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |