Learn More
ACROBiosystems Human Ephrin-A4 / EFNA4 Protein, His Tag
Marque: ACROBiosystems EA4-H5229-1mg
Informations supplémentaires : Poids : 0.00010kg
Description
Ephrin-A4 (EFNA4) is also known as EPH-related receptor tyrosine kinase ligand 4 (LERK4), EPLG4, which belongs to the ephrin family. EFNA4 contains one ephrin RBD (ephrin receptor-binding) domain. EFNA4 is cell surface GPI-bound ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. EFNA4 binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. EFNA4 may play a role in the interaction between activated B-lymphocytes and dendritic cells in tonsils.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.5 kDa. The protein migrates as 18-20 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAI07484 | |
1 mg | |
Leu 26 - Gly 171 | |
EFNA4,EPLG4,LERK4,Ephrin-A4 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
17.5 kDa | |
Human Ephrin-A4, His Tag (EA4-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gly 171 (Accession # AAI07484). | |
-20°C | |
Ephrin-A4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |