Learn More
Abnova™ Human FMN2 Partial ORF (AAH14364.2, 144 a.a. - 243 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00056776-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
Formin homology (FH) domain proteins (e.g., FMN; MIM 136535) play a role in cytoskeletal organization and/or establishment of cell polarity.[supplied by OMIM]
Sequence: DQEAEENSLTETHKCFLETTAYFFMKPKLGEKEVSPNAFFSIWHEFSSDFKDFWKKENKLLLQERVKEAEEVCRQKKGKSLYKIKPRHDSGIKAKISMKTSpécification
AAH14364.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DQEAEENSLTETHKCFLETTAYFFMKPKLGEKEVSPNAFFSIWHEFSSDFKDFWKKENKLLLQERVKEAEEVCRQKKGKSLYKIKPRHDSGIKAKISMKT | |
RUO | |
FMN2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
56776 | |
FMN2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FMN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |