Learn More
Invitrogen™ Human Glucocorticoid Receptor alpha (aa 755-771) Synthetic Peptide
Synthetic Peptide
Marque: Invitrogen™ PEP221
Code nomenclature Nacres: NA.26
Informations supplémentaires : CAS : 69-74-9 Poids : 0.01000kg
Description
Human GR alpha (residues 755-771) Synthetic Peptide, PEP-221, from Invitrogen.
PEP-221 is a 18 amino acid synthetic peptide corresponding to amino acid residues 755-771 of human GR alpha. The sequence of this peptide is (amino to carboxy terminus): C (755) E I I T N Q I P K Y S N G N I K K (771).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-516. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Spécification
2908 | |
purified | |
Human Glucocorticoid Receptor alpha (aa 755-771) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P04150 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
NR3C1 | |
No preservative | |
50 μg | |
C(755) E I I T N Q I P K Y S N G N I K K(771) |