Learn More
ACROBiosystems Human Glutaredoxin 1 / GLRX1 Protein, His Tag
Marque: ACROBiosystems GLX-H5149-100UG
Informations supplémentaires : Poids : 0.00001kg
Description
Glutaredoxin-1 (GRX1) is also known as Thioltransferase-1 (TTase-1), GLRX, GRX, GLRX1, which belongs to the glutaredoxin family. GRX1 / GLRX contains one glutaredoxin domain, which exists in either a reduced or an oxidized form. GRX1 / GLRX has a glutathione-disulfide oxidoreductase activity in the presence of NADPH and glutathione reductase and functions as electron carriers in the glutathione-dependent synthesis of deoxyribonucleotides by the enzymeribonucleotide reductase.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 12.7 kDa. The protein migrates as 12 kDa under reducing (R) condition (SDS-PAGE).Spécification
NP_001112362 | |
100 μg | |
Met 1 - Gln 106 | |
GLRX,GRX,TTase-1,GLRX1,GRX1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
12.7 kDa | |
Human Glutaredoxin 1, His Tag (GLX-H5149) is expressed from E.coli cells. It contains AA Met 1 - Gln 106 (Accession # NP_001112362). | |
-20°C | |
Glutaredoxin 1 | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 with 1 mM DTT | |
E.coli | |
Other Recombinant Proteins |