Learn More
Human HLA-DR Antibody, R&D Systems™
Code produit 30226438
Change view
Click to view available options
Quantité:
100 μg
25 μg
Conditionnement:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30226438 100 μg 100µg
30226439 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30226438 Fournisseur R&D Systems Code fournisseur MAB11555100
Mouse Monoclonal Antibody
HLA-DR Monoclonal antibody specifically detects HLA-DR in Human samples. It is validated for Immunohistochemistry,Western Blot,Simple Western,Multiplex Immunofluorescence
TRUSTED_SUSTAINABILITY
Spécification
Antigène HLA-DR
Applications Immunohistochemistry, Western Blot, Simple Western, Multiplex
Classification Monoclonal
Clone 1083110
Conjugué Unconjugated
Dilution Western Blot 2 μg/mL, Simple Western 20 μg/mL, Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 1 μg/mL
Formule Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Numéro d’ordre du gène P01903
Alias de gène FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLADR, HLA-DRA, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MHC Class II DR, MLRW
Espèces hôtes Mouse
Immunogène Synthetic Peptide, Accession # P01903
Méthode de purification Protein A or G purified from hybridoma culture supernatant
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 3122
Reconstitution Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Espèces cibles Human
Contenu et stockage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Forme Purified
Isotype Not Available