Learn More
ACROBiosystems Human ICAM-1 / CD54 Protein, His Tag
423.38€ - 3108.82€
Spécification
Numéro d’adhésion | AAH15969.1 |
---|---|
Poids moléculaire | 51.4 kDa |
Source | Human ICAM-1, His Tag (IC1-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Glu 480 (Accession # P05362-1). |
Immunogène | Gln 28 - Glu 480 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15866957
|
ACROBiosystems
IC1-H52H5-100UG |
100 μg |
423.38€
100µg |
Expédition estimée: 27-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15876957
|
ACROBiosystems
IC1-H52H5-1MG |
1 mg |
3108.82€
1mg |
Expédition estimée: 27-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Inter-Cellular Adhesion Molecule 1 (ICAM-1) is also known as Cluster of Differentiation 54 (CD54), is a member of the immunoglobulin superfamily, and is a cell surface glycoprotein which is typically expressed in low concentrations on endothelial cells and cells of the immune system. The protein encoded by this gene is a type of intercellular adhesion molecule continuously present in low concentrations in the membranes of leukocytes and endothelial cells. Upon cytokine stimulation, the concentrations greatly increase. ICAM-1 can be induced by interleukin-1 (IL-1) and tumor necrosis factor alpha (TNFα) and is expressed by the vascular endothelium, macrophages, and lymphocytes. ICAM-1 is a ligand for LFA-1 (integrin), a receptor found on leukocytes. When activated, leukocytes bind to endothelial cells via ICAM-1/LFA-1 and then transmigrate into tissues. ICAM-1 has been implicated in subarachnoid hemorrhage (SAH). Levels of ICAM-1 are shown to be significantly elevated in patients with SAH over control subjects in many studies. ICAM-1 expressed by respiratory epithelial cells is also the binding site for rhinovirus, the causative agent of most common colds.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 51.4 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH15969.1 | |
Human ICAM-1, His Tag (IC1-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Glu 480 (Accession # P05362-1). | |
-20°C | |
ICAM-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
51.4 kDa | |
Gln 28 - Glu 480 | |
ICAM1,BB2,CD54,P3.58 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |