Learn More
Human IFN-beta Antibody, R&D Systems™
GREENER_CHOICE
Code produit 18745479
Change view
Click to view available options
Quantité:
100 μg
25 μg
Conditionnement:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18745479 25 μg 25µg
18713609 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18745479 Fournisseur R&D Systems Code fournisseur MAB8141SP
Mouse Monoclonal Antibody
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Neutralization, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Spécification
Antigène IFN-beta
Applications Neutralization, Immunocytochemistry
Classification Monoclonal
Clone 937912
Conjugué Unconjugated
Dilution Neutralization 0.2-1.2 ug/mL, Immunocytochemistry 8-25 ug/mL
Formule Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Alias de gène Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Espèces hôtes Mouse
Immunogène Chinese hamster ovary cell line CHO-derived recombinant human IFN-beta
Méthode de purification Protein A or G purified from hybridoma culture supernatant
Quantité 25 μg
Primaire ou secondaire Primary
Identification génétique (Entrez) 3456
Spécificité du test Detects human IFN-beta in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Espèces cibles Human
Contenu et stockage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Forme Supernatant
Isotype IgG2b