Learn More
Human IFN-beta Antibody, R&D Systems™
GREENER_CHOICE
Code produit 18738878
Change view
Click to view available options
Quantité:
1 mg
100 μg
25 μg
500 ug
Conditionnement:
100µg
1mg
25µg
500µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18738878 25 μg 25µg
18712549 100 μg 100µg
18158800 500 ug 500µg
18121331 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 18738878 Fournisseur R&D Systems Code fournisseur MAB814RSP
Mouse Monoclonal Antibody
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Spécification
Antigène IFN-beta
Applications Neutralization
Classification Monoclonal
Clone 76703R
Conjugué Unconjugated
Dilution Neutralization 7-21 ug/mL
Formule Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Numéro d’ordre du gène P01574
Alias de gène Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Symboles de gène(s) IFNB1
Espèces hôtes Mouse
Immunogène E. coli-derived recombinant human IFN-beta Met22-Asn187 Accession # P01574
Méthode de purification Protein A or G purified from cell culture supernatant
Quantité 25 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 3456
Spécificité du test Detects human IFN-beta in direct ELISAs.
Espèces cibles Human
Contenu et stockage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Forme Purified
Isotype IgG1
Afficher plus Afficher moins
For Research Use Only