Learn More
Human IFN-beta Antibody, R&D Systems™
GREENER_CHOICE
Code produit 18750067
Change view
Click to view available options
Quantité:
100 μg
25 μg
Conditionnement:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18750067 25 μg 25µg
18791447 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18750067 Fournisseur R&D Systems Code fournisseur MAB11127SP
Mouse Monoclonal Antibody
IFN-beta Monoclonal antibody specifically detects IFN-beta in Human samples. It is validated for Neutralization
TRUSTED_SUSTAINABILITY
Spécification
Antigène IFN-beta
Applications Neutralization
Classification Monoclonal
Clone 1020369
Conjugué Unconjugated
Dilution Neutralization
Formule Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Numéro d’ordre du gène P01574
Alias de gène Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Espèces hôtes Mouse
Immunogène CHO-expressed recombinant Human IFN-beta protein, Met22-Asn187, Accession # P01574
Méthode de purification Protein A or G purified from hybridoma culture supernatant
Quantité 25 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 3456
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Espèces cibles Human
Contenu et stockage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Forme Purified
Isotype IgG1