missing translation for 'onlineSavingsMsg'
Learn More
Gibco™ Human IL-29 (IFN lambda-1) Recombinant Protein, PeproTech® Code produit.: 17821713

Gibco™ Human IL-29 (IFN lambda-1) Recombinant Protein, PeproTech®

Code produit. 17821713
5 μg, 5µg
Code nomenclature Nacres: NA.28
Click to view available options
Quantité:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
20µg
250µg
500µg
5µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17821713

Marque: Gibco™ 30002L5UG

Faites de belles économies avec le Laboratoire de la Curiosité
Utilisez le code promo 24110 lors de la validation de la commande
Voir les promotions disponibles

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Recombinant Human IFN-lambda1 is a 19.8 kDa protein containing 178 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IFN-lambdas constitute a novel family of interferons that mediate the induction of anti-viral protection in a wide variety of cells. There are three members in this family, namely lambda1, lambda2, and lambda3, which are also known as IL-29, IL-28A, and IL-28B, respectively. Similar to type I IFNs, IFN-lambdas activate an intracellular signaling to promote gene expression. IFN-lambdas mediate their anti-viral protection through a class II cytokine receptor complex composed of two essential receptor proteins, CRF2-12/IFN-lambdaR1, which is unique to IFN-lambdas, and CFR2-4/IL-10R2, which is shared with IL-10, IL-22, and IL-26 receptors. The IFN-lambdas, IL-28 and IL-29, have recently been reported to prime dendritic cells to induce proliferation of Foxp3-bearing regulatory T cells. DCs that differentiate in response to IFN-lambdas express high levels of class I and II MHC gene products, but low levels of costimulatory molecules. Moreover, these cells can specifically induce IL-2-dependent proliferation of CD4+CD25+FOXP3+ T cells with contact-dependent suppressive activity.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion Q8IU54
À utiliser avec (application) Functional Assay
Formule protein with no preservative
Identification génétique (Entrez) 282618
Poids moléculaire 19.8 kDa
Nom Human IL-29 (IFN lambda-1)
Quantité 5 μg
Source E. coli
État réglementaire RUO
Concentration en endotoxines <1 EU/ μg
Alias de gène cytokine Zcyto21; Cytokine Zcyto22; FNL1; IFN lambda1; IFNL1; IFNL3; IFN-lambda-1; IFN-lambda-3; IFN-lambda-4; IL28B; IL-28 B; IL28C; IL-28 C; IL29; IL-29; ILN; interferon lambda 1; interferon lambda 3; Interferon lambda1; interferon lambda-1; Interferon lambda-3; interferon, lambda 1; interferon, lambda 3; interferon, lambda 4; interferon-lambda-1; Interleukin; interleukin 29 (interferon, lambda 1); interleukin-28 B; Interleukin-28 C; interleukin-29; ZCYTO21; ZCYTO22
Nom usuel IL-29
Symbole de gène(s) IFNL1
Activité biologique Determined by its ability to activate STAT phosphorylation in an ISRE Luciferase Reporter Assay using human colon carcinoma COLO205 cells. The expected ED50 is 0.2-0.5 ng/ml
Conjugué Unconjugated
Recombinant Recombinant
Séquence PTSKPTTTGK GCHIGRFKSL SPQELASFKK ARDALEESLK LKNWSCSSPV FPGNWDLRLL QVRERPVALE AELALTLKVL EAAAGPALED VLDQPLHTLH HILSQLQACI QPQPTAGPRP RGRLHHWLHR LQEAPKKESA GCLEASVTFN LFRLLTRDLK YVADGNLCLR TSTHPEST
Contenu et stockage -20°C
Système d’expression E. coli
Forme Lyophilized
Pureté ou qualité ≥ 98% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
Gibco™ Human IL-29 (IFN lambda-1) Recombinant Protein, PeproTech® >

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis