missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IMP5 Partial ORF (NP_787078, 29 a.a. - 122 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00162540-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
Sequence: VVSENWSKDYCILFSSDYITLPRDLHHAPLLPLYDGTKAPWCPGEDSPHQAQLRSPSQRPLRQTTAMVMRGNCSFHTKGWLAQGQGAHGLLIVSSpécification
NP_787078 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VVSENWSKDYCILFSSDYITLPRDLHHAPLLPLYDGTKAPWCPGEDSPHQAQLRSPSQRPLRQTTAMVMRGNCSFHTKGWLAQGQGAHGLLIVS | |
RUO | |
IMP5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
162540 | |
IMP5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SPPL2c | |
IMP5 | |
Recombinant | |
wheat germ expression system |