missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human JPH3 Synthetic Peptide
Synthetic Peptide
Marque: Invitrogen™ PEP0763
Code nomenclature Nacres: NA.26
Informations supplémentaires : Poids : 0.01000kg
Description
JPH3 Synthetic Peptide, PEP-0763, from Invitrogen.
This peptide corresponds to 18 amino acids near the carboxy terminus of human JPH3.PEP-0763 can be used as a blocking peptide with polyclonal antibody PA5-20643.Spécification
57338 | |
purified | |
Human JPH3 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q8WXH2 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
JPH3 | |
0.02% sodium azide | |
50 μg | |
PBS |