missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LRFN4 Synthetic Peptide
Synthetic Peptide
Marque: Invitrogen™ PEP0814
Code nomenclature Nacres: NA.26
Informations supplémentaires : Poids : 0.01000kg
Description
LRFN4 Synthetic Peptide, PEP-0814, from Invitrogen.
This peptide corresponds to 14 amino acids near the carboxy terminus of human LRFN4.PEP-0814 can be used as a blocking peptide with polyclonal antibody PA5-20699.Spécification
78999 | |
purified | |
Human LRFN4 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q6PJG9 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
LRFN4 | |
0.02% sodium azide | |
50 μg | |
PBS |