Learn More
ACROBiosystems Human LRRC4 Protein, His Tag
Marque: ACROBiosystems LR4-H5221-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
LRRC4 (Leu-rich repeat/LRR-containing glycoprotein 4), LRRC4B and LRRC4C are post-synaptic adhesion molecules of the LRR protein family that induce excitatory synapse formation. LRRC4 is also known as Brain tumor-associated protein BAG, Netrin-G2 ligand (NGL-2), Nasopharyngeal carcinoma-associated gene 14 protein (NAG14), which contains 1 Ig-like (immunoglobulin-like) domain and 9 LRR (leucine-rich) repeats, 1 LRRCT domain, 1 LRRNT domain. LRRC4 / NGL-2 specifically expressed in brain. LRRC4 / NGL-2 regulates the formation of exitatory synapses through the recruitment of pre-and-postsynaptic proteins. LRRC4 / NGL-2 organize the lamina/pathway-specific differentiation of dendrites. LRRC4 / NGL-2 plays a important role for auditory synaptic responses and involved in the suppression of glioma.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.8 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAI11562.1 | |
1 mg | |
Ala 39 - Lys 527 | |
LRRC4,NGL-2,BAG,NAG14 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
55.8 kDa | |
Human LRRC4, His Tag (LR4-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 39 - Lys 527 (Accession # AAI11562.1). | |
-20°C | |
LRRC4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |