Learn More
Abnova™ Human MAGI2 Partial ORF (NP_036433, 519 a.a. - 628 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00009863-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene interacts with atrophin-1. Atrophin-1 contains a polyglutamine repeat, expansion of which is responsible for dentatorubral and pallidoluysian atrophy. This encoded protein is characterized by two WW domains, a guanylate kinase-like domain, and multiple PDZ domains. It has structural similarity to the membrane-associated guanylate kinase homologue (MAGUK) family. [provided by RefSeq]
Sequence: PANSMVPPLAIMERPPPVMVNGRHNYETYLEYISRTSQSVPDITDRPPHSLHSMPTDGQLDGTYPPPVHDDNVSMASSGATQAELMTLTIVKGAQGFGFTIADSPTGQRVSpécification
NP_036433 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PANSMVPPLAIMERPPPVMVNGRHNYETYLEYISRTSQSVPDITDRPPHSLHSMPTDGQLDGTYPPPVHDDNVSMASSGATQAELMTLTIVKGAQGFGFTIADSPTGQRV | |
RUO | |
MAGI2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9863 | |
MAGI2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACVRIP1/AIP1/ARIP1/MAGI-2/SSCAM | |
MAGI2 | |
Recombinant | |
wheat germ expression system |