missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAP3K15 Partial ORF (NP_001001671, 691 a.a. - 786 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00389840-Q01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
Sequence: LYHLQLKLKSNCITENPAGPYGQRTDKELIDWLRLQGADAKTIEKIVEEGYTLSDILNEITKEDLRYLRLRGGLLCRLWSAVSQYRRAQEASETKDSpécification
NP_001001671 | |
Liquid | |
389840 | |
MAP3K15 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ16518/bA723P2.3 | |
MAP3K15 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.3kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LYHLQLKLKSNCITENPAGPYGQRTDKELIDWLRLQGADAKTIEKIVEEGYTLSDILNEITKEDLRYLRLRGGLLCRLWSAVSQYRRAQEASETKD | |
RUO | |
MAP3K15 | |
Wheat Germ (in vitro) | |
GST |