Learn More
Abnova™ Human MAP3K3 Partial ORF (AAH10464.1, 1 a.a. - 89 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Spécification
Numéro d’adhésion | AAH10464.1 |
---|---|
À utiliser avec (application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Identification génétique (Entrez) | 4215 |
Poids moléculaire | 35.53kDa |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16151155
|
Abnova™
H00004215-Q01.10UG |
10 ug |
335.00€
10µg |
Expédition estimée: 18-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16161155
|
Abnova™
H00004215-Q01.25UG |
25 ug |
508.00€
25µg |
Expédition estimée: 18-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This gene product is a 626-amino acid polypeptide that is 96.5% identical to mouse Mekk3. Its catalytic domain is closely related to those of several other kinases, including mouse Mekk2, tobacco NPK, and yeast Ste11. Northern blot analysis revealed a 4.6-kb transcript that appears to be ubiquitously expressed. This protein directly regulates the stress-activated protein kinase (SAPK) and extracellular signal-regulated protein kinase (ERK) pathways by activating SEK and MEK1/2 respectively; it does not regulate the p38 pathway. In cotransfection assays, it enhanced transcription from a nuclear factor kappa-B (NFKB)-dependent reporter gene, consistent with a role in the SAPK pathway. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: MNEANVMLPYSGKEEPVLPVAMTLPLPGRGPRCGTAATEGGSSFVNAVVSVLQVGVTLMLYPVSKLETVCALWALSTPALGLGLGCIEKSpécification
AAH10464.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAPKKK3/MEKK3 | |
MAP3K3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4215 | |
MAP3K3 (Human) Recombinant Protein (Q01) | |
MNEANVMLPYSGKEEPVLPVAMTLPLPGRGPRCGTAATEGGSSFVNAVVSVLQVGVTLMLYPVSKLETVCALWALSTPALGLGLGCIEK | |
RUO | |
MAP3K3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |