Learn More
ACROBiosystems Human Mesothelin / MSLN (37-286) Protein, His Tag
Marque: ACROBiosystems MSN-H5222-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
Mesothelin (MSLN) is also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, which belongs to the mesothelin family. Mesothelin / MSLN can be proteolytically cleaved into the following two chains by a furin-like convertase: Megakaryocyte-potentiating factor (MPF) and the cleaved form of mesothelin. Both MPF and the cleaved form of mesothelin are N-glycosylated. Mesothelin / MSLN can interacts with MUC16. The membrane-anchored forms of MSLN may play a role in cellular adhesion. MPF potentiates megakaryocyte colony formation in vitro.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.5 kDa. The protein migrates as 32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH09272 | |
1 mg | |
Leu 37 - Arg 286 | |
MSLN,Mesothelin,MPF | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
27.5 kDa | |
Human Mesothelin (37-286), His Tag (MSN-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 37 - Arg 286 (Accession # AAH09272). | |
-20°C | |
Mesothelin | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |