missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human METAP2 (aa 10-101) Control Fragment Recombinant Protein
Code nomenclature Nacres: NA.28
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The ADP-ribosylation factor (Arf) family are highly conserved members of the Ras superfamily of regulatory GTP-binding proteins. Arf proteins participate in routing of intracellular proteins to and within the Golgi complex. Cellular functions include maintenance of organelle integrity, coat protein assembly, as an activator of phospholipase D. The Arf family is divided functionally into the Arf and the Arf-like (Arl) proteins. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6) and members of each class share a common gene organization.
Spécification
Spécification
| Numéro d’adhésion | P50579 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 10988 |
| Nom | Human METAP2 (aa 10-101) Control Fragment |
| Quantité | 100 μl |
| État réglementaire | RUO |
| Alias de gène | 4930584B20Rik; A930035J23Rik; AI047573; AL024412; Amp2; AU014659; eIF-2-associated p67; eIF-2-associated p67 homolog; initiation factor 2 associated 67 kDa protein; initiation factor 2-associated 67 kDa glycoprotein; initiation factor 2-associated protein (p67); MAP 2; MAP2; metAP 2; Metap2; Methionine aminopeptidase 2; methionyl aminopeptidase 2; Mnpep; p67; p67eIF2; Peptidase M; peptidase M 2; similar to methionyl aminopeptidase 2; testicular tissue protein Li 17 |
| Nom usuel | METAP2 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Invitrogen™ Human METAP2 (aa 10-101) Control Fragment Recombinant Protein >
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu