missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MGST3 Partial ORF (NP_004519, 28 a.a. - 84 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00004259-Q01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16151245

  • 335.00€ / 10µg
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The MAPEG (Membrane Associated Proteins in Eicosanoid and Glutathione metabolism) family consists of six human proteins, several of which are involved the production of leukotrienes and prostaglandin E, important mediators of inflammation. This gene encodes an enzyme which catalyzes the conjugation of leukotriene A4 and reduced glutathione to produce leukotriene C4. This enzyme also demonstrates glutathione-dependent peroxidase activity towards lipid hydroperoxides. [provided by RefSeq]

Sequence: NVSKARKKYKVEYPIMYSTDPENGHIFNCIQRAHQNTLEVYPPFLFFLAVGGVYHPR
Spécification

Spécification

NP_004519
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.01kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NVSKARKKYKVEYPIMYSTDPENGHIFNCIQRAHQNTLEVYPPFLFFLAVGGVYHPR
RUO
MGST3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4259
MGST3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GST-III
MGST3
Recombinant
wheat germ expression system
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions