missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Doublecortin/DCX Antibody, R&D Systems™

Description
Doublecortin specifically detects Doublecortin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
| Antigène | Doublecortin |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 8-25 μg/mL |
| Formule | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Numéro d’ordre du gène | O43602 |
| Alias de gène | DBCN, Doublecortex, doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, LISX, X-linked (doublecortin) |
| Espèces hôtes | Sheep |
| Immunogène | E.coli-derived recombinant human Doublecortin/DCX, Met116-Thr293, Accession # O43602 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?