Learn More
Abnova™ Human NIPSNAP1 Full-length ORF (AAH02371.1, 1 a.a. - 284 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00008508-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
NIPSNAP1 protein has a strong sequence similarity to the central portion of a hypothetical protein encoded by C. elegans chromosome III between a 4-nitrophenylphosphatase (NIP) domain and non-neuronal SNAP25-like protein. The NIPSNAP1 gene is located between the NF2 and pK1.3 genes on 22q12. [provided by RefSeq]
Sequence: MAPRLCSISVTARRLLGGPGPRAGDVASAAAARFYSKDNEGSWFRSLFVHKVDPRKDAHSTLLSKKETSNLYKIQFHNVKPEYLDAYNSLTEAVLPKLHLDEDYPCSLVGNWNTWYGEQDQAVHLWRFSGGYPALMDCMNKLKNNKEYLEFRRERSQMLLSRRNQLLLEFSFWNEPQPRMGPNIYELRTYKLKPGTMIEWGNNWARAIKYRQENQEAVGGFFSQIGELYVVHHLWAYKDLQSREETRNAAWRKRGWDENVYYTVPLVRHMESRIMIPLKISPLQSpécification
AAH02371.1 | |
Liquid | |
8508 | |
NIPSNAP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAPRLCSISVTARRLLGGPGPRAGDVASAAAARFYSKDNEGSWFRSLFVHKVDPRKDAHSTLLSKKETSNLYKIQFHNVKPEYLDAYNSLTEAVLPKLHLDEDYPCSLVGNWNTWYGEQDQAVHLWRFSGGYPALMDCMNKLKNNKEYLEFRRERSQMLLSRRNQLLLEFSFWNEPQPRMGPNIYELRTYKLKPGTMIEWGNNWARAIKYRQENQEAVGGFFSQIGELYVVHHLWAYKDLQSREETRNAAWRKRGWDENVYYTVPLVRHMESRIMIPLKISPLQ | |
RUO | |
NIPSNAP1 | |
Yes | |
wheat germ expression system |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NIPSNAP1 | |
Wheat Germ (in vitro) | |
GST |