missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human NT-4, Animal-Free Recombinant Protein, PeproTech®

E. coli Recombinant Protein

Marque:  Gibco™ AF-450-04-1MG

Code nomenclature Nacres: NA.75

Informations supplémentaires : Poids : 0.16500kg

 Afficher plus de versions de ce produit

Code produit. 17893073

  • 5650.00€ / 1mg
Expédition estimée: 12-08-2024
pour voir le stock disponible 

Description

Description

This gene is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons. The expression of this gene is ubiquitous and less influenced by environmental signals. While knock-outs of other neurotrophins including nerve growth factor, brain-derived neurotrophic factor, and neurotrophin 3 prove lethal during early postnatal development, NTF5-deficient mice only show minor cellular deficits and develop normally to adulthood.
Spécification

Spécification

P34130
protein with no preservative
28.1
1 mg
-20°C
<0.1 EU/μg
NTF4
Unconjugated
GVSETAPASR RGELAVCDAV SGWVTDRRTA VDLRGREVEV LGEVPAAGGS PLRQYFFETR CKADNAEEGG PGAGGGGCRG VDRRHWVSEC KAKQSYVRAL TADAQGRVGW RWIRIDTACV CTLLSRTGRA
≥ 98% by SDS-PAGE gel and HPLC analyses.
ELISA, Functional Assay
4909
Human NT-4, Animal-Free
E. coli
RUO
2900040K06Rik; AI462899; GLC10; GLC1O; neurotrophic factor 4; neurotrophic factor 5; neurotrophin 4; Neurotrophin 4/5; neurotrophin 5; Neurotrophin 5 (neurotrophin 4/5); neurotrophin 5 (neurotrophin 4/5); neurotrophin-4; Neurotrophin-5; neutrophic factor 4; Neutrophic factor 5; NT4; NT-4; NT4/5; NT-4/5; NT4/NT5; NT4P; NT5; NT-5; Ntf4; Ntf5; Ntf-5
Human
Recombinant
Lyophilized
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions