missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human OCC1 Synthetic Peptide
Synthetic Peptide
Marque: Invitrogen™ PEP0791
Code nomenclature Nacres: NA.26
Informations supplémentaires : Poids : 0.01000kg
Description
OCC-1 Synthetic Peptide, PEP-0791, from Invitrogen.
This peptide corresponds to 16 amino acids near the amino terminus of human OCC-1PEP-0791 can be used as a blocking peptide with polyclonal antibody PA5-20673.Spécification
387882 | |
purified | |
Human OCC1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q8TAD7 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
C12orf75 | |
0.02% sodium azide | |
50 μg | |
PBS |