missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OTOP2 Partial ORF (NP_835454, 423 a.a. - 493 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00092736-Q01.10ug
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 0.00010kg
Description
Sequence: ESLHRGPPGAEPHSTHPKEPCQDLTFTNLDALHTLSACPPNPGLVSPSPSDQREAVAIVSTPRSQWRRQCLSpécification
NP_835454 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.55kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ESLHRGPPGAEPHSTHPKEPCQDLTFTNLDALHTLSACPPNPGLVSPSPSDQREAVAIVSTPRSQWRRQCL | |
RUO | |
OTOP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
92736 | |
OTOP2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
OTOP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |