missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human P2RX5 Full-length ORF (NP_002552.2, 1 a.a. - 422 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Spécification
Numéro d’adhésion | NP_002552.2 |
---|---|
À utiliser avec (application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Identification génétique (Entrez) | 5026 |
Poids moléculaire | 73.6kDa |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16152285
|
Abnova™
H00005026-P01.25UG |
25 ug |
508.00€
25µg |
Expédition estimée: 20-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16142285
|
Abnova™
H00005026-P01.10UG |
10 ug |
335.00€
10µg |
Expédition estimée: 20-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel. Several characteristic motifs of ATP-gated channels are present in its primary structure, but, unlike other members of the purinoceptors family, this receptor has only a single transmembrane domain. Three transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MGQAGCKGLCLSLFDYKTEKYVIAKNKKVGLLYRLLQASILAYLVVWVFLIKKGYQDVDTSLQSAVITKVKGVAFTNTSDLGQRIWDVADYVIPAQGENVFFVVTNLIVTPNQRQNVCAENEGIPDGACSKDSDCHAGEAVTAGNGVKTGRCLRRENLARGTCEIFAWCPLETSSRPEEPFLKEAEDFTIFIKNHIRFPKFNFSKSNVMDVKDRSFLKSCHFGPKNHYCPIFRLGSVIRWAGSDFQDIALEGGVIGINIEWNCDLDKAASECHPHYSFSRLDNKLSKSVSSGYNFRFARYYRDAAGVEFRTLMKAYGIRFDVMVNGKGAFFCDLVLIYLIKKREFYRDKKYEEVRGLEDSSQEAEDEASGLGLSEQLTSGPGLLGMPEQQELQEPPEAKRGSSSQKGNGSVCPQLLEPHRSTSpécification
NP_002552.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGQAGCKGLCLSLFDYKTEKYVIAKNKKVGLLYRLLQASILAYLVVWVFLIKKGYQDVDTSLQSAVITKVKGVAFTNTSDLGQRIWDVADYVIPAQGENVFFVVTNLIVTPNQRQNVCAENEGIPDGACSKDSDCHAGEAVTAGNGVKTGRCLRRENLARGTCEIFAWCPLETSSRPEEPFLKEAEDFTIFIKNHIRFPKFNFSKSNVMDVKDRSFLKSCHFGPKNHYCPIFRLGSVIRWAGSDFQDIALEGGVIGINIEWNCDLDKAASECHPHYSFSRLDNKLSKSVSSGYNFRFARYYRDAAGVEFRTLMKAYGIRFDVMVNGKGAFFCDLVLIYLIKKREFYRDKKYEEVRGLEDSSQEAEDEASGLGLSEQLTSGPGLLGMPEQQELQEPPEAKRGSSSQKGNGSVCPQLLEPHRST | |
RUO | |
P2RX5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
5026 | |
P2RX5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC47755/P2X5/P2X5R | |
P2RX5 | |
Recombinant | |
wheat germ expression system |