Learn More
ACROBiosystems Human PD-1 / PDCD1 Protein, Fc Tag, low endotoxin
MALS verified
Marque:  ACROBiosystems PD1-H5257-100UG
Informations supplémentaires : Poids : 0.00001kg
 Afficher plus de versions de ce produit
Code produit. 15877457
  • 399.19€ / 100µg
Expédition estimée: 18-06-2024
pour voir le stock disponible 
Description
Description
Programmed cell death protein 1 (PD-1) is also known as CD279 and PDCD1, is a type I membrane protein and is a member of the extended CD28/CTLA-4 family of T cell regulators. PDCD1 is expressed on the surface of activated T cells, B cells, macrophages, myeloid cells and a subset of thymocytes. PD-1 has two ligands, PD-L1 and PD-L2, which are members of the B7 family. PD-L1 is expressed on almost all murine tumor cell lines, including PA1 myeloma, P815 mastocytoma, and B16 melanoma upon treatment with IFN-γ. PD-L2 expression is more restricted and is expressed mainly by DCs and a few tumor lines. PD1 inhibits the T-cell proliferation and production of related cytokines including IL-1, IL-4, IL-10 and IFN-γ by suppressing the activation and transduction of PI3K/AKT pathway. In addition, coligation of PD1 inhibits BCR-mediating signal by dephosphorylating key signal transducer. In vitro, treatment of anti-CD3 stimulated T cells with PD-L1-Ig results in reduced T cell proliferation and IFN-γ secretion. Monoclonal antibodies targeting PD-1 that boost the immune system are being developed for the treatment of cancer.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.1 kDa. The protein migrates as 56-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification
Spécification
NP_005009.2
100 μg
Leu 25 - Gln 167
PDCD1,PD1,CD279,SLEB2
Unconjugated
Fc Tag
Less than 0.1 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
42.1 kDa
Human PD-1, Fc Tag, low endotoxin (PD1-H5257) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # NP_005009.2).
-20°C
PD-1
Human
PBS, pH7.4
HEK293
Immune Checkpoint Proteins