Learn More
ACROBiosystems Human PD-L1 / B7-H1 Protein, His Tag
MALS verified
Marque: ACROBiosystems PD1-H5229-1MG
Informations supplémentaires : Poids : 0.00001kg
Description
Programmed cell death 1 ligand 1 (PD-L1) is also known as cluster of differentiation (CD274) or B7 homolog 1 (B7-H1), is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses. B7-H1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. PD-L1 is highly expressed in the heart, skeletal muscle, placenta and lung and weakly expressed in the thymus, spleen, kidney and liver. PD-L1 is expressed on activated T-cells, B-cells, dendritic cells, keratinocytes and monocytes. PD-L1 is up-regulated on T- and B-cells, dendritic cells, keratinocytes and monocytes after LPS and IFNG activation and up-regulated in B-cells activated by surface Ig cross-linking. PD-L1 involve in the costimulatory signal, essential for T-cell proliferation and production of IL10 and IFNG, in an IL2-dependent and a PDCD1-independent manner.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.0 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_054862.1 | |
1 mg | |
Phe 19 - Arg 238 | |
PD-L1,CD274,B7-H1,PDCD1L1,PDCD1LG1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>98% as determined by SDS-PAGE |
26.0 kDa | |
Human PD-L1, His Tag (PD1-H5229) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # NP_054862.1). | |
-20°C | |
PD-L1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |