Learn More
ACROBiosystems Human PD-L1 / B7-H1 Protein, Llama IgG2b Fc Tag, low endotoxin
low endotoxin
423.38€ - 3108.82€
Spécification
Numéro d’adhésion | NP 054862.1 |
---|---|
Poids moléculaire | 53.0 kDa |
Source | Human PD-L1, Llama IgG2b Fc Tag, low endotoxin (PDL-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19- Arg 238 (Accession # Q9NZQ7-1). |
Immunogène | Phe 19- Arg 238 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15887527
|
ACROBiosystems
PDL-H5250-1MG |
1 mg |
3108.82€
1mg |
Expédition estimée: 10-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15877527
|
ACROBiosystems
PDL-H5250-100UG |
100 μg |
423.38€
100µg |
Expédition estimée: 10-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Programmed cell death 1 ligand 1 (PDL1) is also known as B7-H, B7H1, MGC142294, MGC142296, PD-L1, PDCD1L1 and PDCD1LG1,which is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses.PDL1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. This protein is broadly expressed in the majority of peripheral tissues as well as hematopoietic cells. Interaction between PDL1 and its receptors belonging to the CD28 family of molecules provide both stimulatory and inhibitory signals in regulating T cell activation and tolerance. PDL1 may inhibit ongoing T-cell responses by inducing apoptosis and arresting cell-cycle progression.
This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 53.0 kDa. The protein migrates as 65-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP 054862.1 | |
Human PD-L1, Llama IgG2b Fc Tag, low endotoxin (PDL-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19- Arg 238 (Accession # Q9NZQ7-1). | |
-20°C | |
PD-L1 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
53.0 kDa | |
Phe 19- Arg 238 | |
PD-L1,CD274,B7-H1,PDCD1L1,PDCD1LG1 | |
Unconjugated | |
Llama IgG Fc Tag | |
Less than 0.01 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |