missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PFKFB4 Full-length ORF (NP_004558.1, 1 a.a. - 469 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00005210-P01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16151238

  • 359.00€ / 10µg
Expédition estimée: 12-07-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Sequence: MASPRELTQNPLKKIWMPYSNGRPALHACQRGVCMTNCPTLIVMVGLPARGKTYISKKLTRYLNWIGVPTREFNVGQYRRDVVKTYKSFEFFLPDNEEGLKIRKQCALAALRDVRRFLSEEGGHVAVFDATNTTRERRATIFNFGEQNGYKTFFVESICVDPEVIAANIVQVKLGSPDYVNRDSDEATEDFMRRIECYENSYESLDEDLDRDLSYIKIMDVGQSYVVNRVADHIQSRIVYYLMNIHVTPRSIYLCRHGESELNLKGRIGGDPGLSPRGREFAKSLAQFISDQNIKDLKVWTSQMKRTIQTAEALGVPYEQWKVLNEIDAGVCEEMTYEEIQDNYPLEFALRDQDKYRYRYPKGESYEDLVQRLEPVIMELERQENVLVICHQAVMRCLLAYFLDKAAEQLPYLKCPLHTVLKLTPVAYGCKVESIFLNVAAVNTHRDRPQNVDISRPPEEALVTVPAHQ
Spécification

Spécification

NP_004558.1
Liquid
5210
PFKFB4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASPRELTQNPLKKIWMPYSNGRPALHACQRGVCMTNCPTLIVMVGLPARGKTYISKKLTRYLNWIGVPTREFNVGQYRRDVVKTYKSFEFFLPDNEEGLKIRKQCALAALRDVRRFLSEEGGHVAVFDATNTTRERRATIFNFGEQNGYKTFFVESICVDPEVIAANIVQVKLGSPDYVNRDSDEATEDFMRRIECYENSYESLDEDLDRDLSYIKIMDVGQSYVVNRVADHIQSRIVYYLMNIHVTPRSIYLCRHGESELNLKGRIGGDPGLSPRGREFAKSLAQFISDQNIKDLKVWTSQMKRTIQTAEALGVPYEQWKVLNEIDAGVCEEMTYEEIQDNYPLEFALRDQDKYRYRYPKGESYEDLVQRLEPVIMELERQENVLVICHQAVMRCLLAYFLDKAAEQLPYLKCPLHTVLKLTPVAYGCKVESIFLNVAAVNTHRDRPQNVDISRPPEEALVTVPAHQ
RUO
PFKFB4
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
80.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PFKFB4
Wheat Germ (in vitro)
GST
Vidéos
FDS
Documents
Certificats
Promotions

Promotions