Learn More
Abnova™ Human PIGC Partial ORF (NP_002633, 1 a.a. - 54 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00005279-Q01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes an endoplasmic reticulum associated protein that is involved in glycosylphosphatidylinositol (GPI) lipid anchor biosynthesis. The GPI lipid anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. The encoded protein is one subunit of the GPI N-acetylglucosaminyl (GlcNAc) transferase that transfers GlcNAc to phosphatidylinositol (PI) on the cytoplasmic side of the endoplasmic reticulum. Two alternatively spliced transcripts that encode the same protein have been found for this gene. A pseudogene on chromosome 11 has also been characterized. [provided by RefSeq]
Sequence: MYAQPVTNTKEVKWQKVLYERQPFPDNYVDRRFLEELRKNIHARKYQYWAVVFESpécification
NP_002633 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.68kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MYAQPVTNTKEVKWQKVLYERQPFPDNYVDRRFLEELRKNIHARKYQYWAVVFE | |
RUO | |
PIGC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5279 | |
PIGC (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GPI2/MGC2049 | |
PIGC | |
Recombinant | |
wheat germ expression system |