missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human pIgR Protein, His Tag
302.42€ - 1935.44€
Spécification
Numéro d’adhésion | AAI10495.1 |
---|---|
Poids moléculaire | 68.7 kDa |
Source | Human pIgR, His Tag (PIR-H5222) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Arg 638 (Accession # AAI10495.1). |
Immunogène | Lys 19 - Arg 638 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15867537
|
ACROBiosystems
PIR-H5222-100UG |
100 μg |
302.42€
100µg |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15877537
|
ACROBiosystems
PIR-H5222-1MG |
1 mg |
1935.44€
1mg |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Polymeric immunoglobulin receptor (PIGR) is also known as Poly-Ig receptor, which can be cleaved into membrane secretory component (SC). PIGR is a type I transmembrane glycoprotein or a member of the immunoglobulin superfamily, and is synthesized by secretory epithelial cells and delivered to the basolateral plasma membrane where it can bind larger polymers of IgA (pIgA ) and pentameric IgM. The complex is then transported across the cell to be secreted at the apical surface. During this process a cleavage occurs that separates the extracellular (known as the secretory component) from the transmembrane segment.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 68.7 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAI10495.1 | |
Human pIgR, His Tag (PIR-H5222) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Arg 638 (Accession # AAI10495.1). | |
-20°C | |
pIgR | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
68.7 kDa | |
Lys 19 - Arg 638 | |
PIGR,Poly-Ig receptor | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |