missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PNO1 Full-length ORF (NP_064528.1, 1 a.a. - 252 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00056902-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
Sequence: MESEMETQSARAEEGFTQVTRKGGRRAKKRQAEQLSAAGEGGDAGRMDTEEARPAKRPVFPPLCGDGLLSGKEETRKIPVPANRYTPLKENWMKIFTPIVEHLGLQIRFNLKSRNVEIRTCKETKDVSALTKAADFVKAFILGFQVEDALALIRLDDLFLESFEITDVKPLKGDHLSRAIGRIAGKGGKTKFTIENVTRTRIVLADVKVHILGSFQNIKMARTAICNLILGNPPSKVYGNIRAVASRSADRFSpécification
NP_064528.1 | |
Liquid | |
56902 | |
PNO1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MESEMETQSARAEEGFTQVTRKGGRRAKKRQAEQLSAAGEGGDAGRMDTEEARPAKRPVFPPLCGDGLLSGKEETRKIPVPANRYTPLKENWMKIFTPIVEHLGLQIRFNLKSRNVEIRTCKETKDVSALTKAADFVKAFILGFQVEDALALIRLDDLFLESFEITDVKPLKGDHLSRAIGRIAGKGGKTKFTIENVTRTRIVLADVKVHILGSFQNIKMARTAICNLILGNPPSKVYGNIRAVASRSADRF | |
RUO | |
PNO1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KHRBP1 | |
PNO1 | |
Yes | |
wheat germ expression system |