Learn More
ACROBiosystems Human Prolactin R / PRLR Protein, Fc Tag
Marque: ACROBiosystems
Informations supplémentaires : Poids : 0.00001kg
Description
As a transmembrane receptor, the prolactin receptor (PRL-R) interacts with prolactin. In addition, the PRL-R also binds and is activated by growth hormone (GH) and human placental lactogen (hPL). Preclinical investigations, epidemiological studies and analyses of tissue specimens from patients strongly support the contribution of prolactin receptor (PRLR) signaling to breast and prostate tumorigenesis and cancer progression. Moreover, The PRLR has been found to be essential for lobuloalveolar maturation of the mammary glands during pregnancy, as evidenced by the fact that PRLR knockout mice show severely impaired development of lobuloalveolar structures.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.0 kDa. The protein migrates as 64 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_000940.1 | |
1 mg | |
Gln 25 - Asp 234 | |
Prolactin R,PRLR,PRL-R | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
51.0 kDa | |
Human Prolactin R, Fc Tag (PRP-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Asp 234 (Accession # P16471-1). | |
-20°C | |
Prolactin R | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Cytokines and Growth Factors |