Learn More
Abnova™ Human PTK9L Partial ORF (AAH00327, 131 a.a. - 230 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00011344-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene was identified by its interaction with the catalytic domain of protein kinase C-zeta. The encoded protein contains an actin-binding site and an ATP-binding site. It is most closely related to twinfilin (PTK9), a conserved actin monomer-binding protein. [provided by RefSeq]
Sequence: FAGYQKHLSSCAAPAPLTSAERELQQIRINEVKTEISVESKHQTLQGLAFPLQPEAQRALQQLKQKMVNYIQMKLDLERETIELVHTEPTDVAQLPSRVPSpécification
AAH00327 | |
Liquid | |
11344 | |
PTK9L (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
A6RP/A6r/MSTP011/PTK9L | |
TWF2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FAGYQKHLSSCAAPAPLTSAERELQQIRINEVKTEISVESKHQTLQGLAFPLQPEAQRALQQLKQKMVNYIQMKLDLERETIELVHTEPTDVAQLPSRVP | |
RUO | |
TWF2 | |
Wheat Germ (in vitro) | |
GST |