Learn More
Abnova™ Human RGS16 Full-length ORF (NP_002919.2, 1 a.a. - 202 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00006004-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene belongs to the 'regulator of G protein signaling' family. It inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits. It also may play a role in regulating the kinetics of signaling in the phototransduction cascade. [provided by RefSeq]
Sequence: MCRTLAAFPTTCLERAKEFKTRLGIFLHKSELGCDTGSTGKFEWGSKHSKENRNFSEDVLGWRESFDLLLSSKNGVAAFHAFLKTEFSEENLEFWLACEEFKKIRSATKLASRAHQIFEEFICSEAPKEVNIDHETRELTRMNLQTATATCFDAAQGKTRTLMEKDSYPRFLKSPAYRDLAAQASAASATLSSCSLDEPSHTSpécification
NP_002919.2 | |
Liquid | |
6004 | |
RGS16 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MCRTLAAFPTTCLERAKEFKTRLGIFLHKSELGCDTGSTGKFEWGSKHSKENRNFSEDVLGWRESFDLLLSSKNGVAAFHAFLKTEFSEENLEFWLACEEFKKIRSATKLASRAHQIFEEFICSEAPKEVNIDHETRELTRMNLQTATATCFDAAQGKTRTLMEKDSYPRFLKSPAYRDLAAQASAASATLSSCSLDEPSHT | |
RUO | |
RGS16 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
A28-RGS14/A28-RGS14P/RGS-R | |
RGS16 | |
Yes | |
wheat germ expression system |