missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human ROR2 / NTRKR2 Protein, His Tag
447.57€ - 3387.03€
Spécification
Numéro d’adhésion | NP_004551.1 |
---|---|
Poids moléculaire | 43.2 kDa |
Source | Human ROR2, His Tag (RO2-H52E5) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # NP_004551.1). |
Immunogène | Glu 34 - Gly 403 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15857607
|
ACROBiosystems
RO2-H52E5-1MG |
1 mg |
3387.03€
1mg |
Expédition estimée: 23-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15867607
|
ACROBiosystems
RO2-H52E5-50UG |
50 μg |
447.57€
50µg |
Expédition estimée: 23-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Tyrosine-protein kinase transmembrane receptor ROR2 is also known as Neurotrophic tyrosine kinase, receptor-related 2 (NTRKR2), which belongs to the protein kinase superfamily and Tyr protein kinase family and ROR subfamily. ROR2 is a homodimer protein, which can binds YWHAB or interact with WTIP. ROR2 may be involved in the early formation of the chondrocytes. It seems to be required for cartilage and growth plate development.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 43.2 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_004551.1 | |
Human ROR2, His Tag (RO2-H52E5) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # NP_004551.1). | |
-20°C | |
ROR2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
43.2 kDa | |
Glu 34 - Gly 403 | |
ROR2,NTRKR2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |