Learn More
ACROBiosystems Human Serpin A1 Protein, His Tag
Marque: ACROBiosystems SE1-H5226-50UG
Informations supplémentaires : Poids : 0.00001kg
Description
Serpin A1 is also known as Alpha-1-antitrypsin (A1AT), serum trypsin inhibitor, alpha-1 proteinase inhibitor (A1PI), AAT, which belongs to the serpin family. Most serpins inactivate enzymes by binding to them covalently, requiring very high levels to perform their function. Like all serine protease inhibitors, A1AT has a characteristic secondary structure of beta sheets and alpha helices. Serpin A1 / A1AT is inhibitor of serine proteases. Its primary target is elastase, but it also has a moderate affinity for plasmin and thrombin. Irreversibly inhibits trypsin, chymotrypsin and plasminogen activator. The aberrant form of SerpinA1 inhibits insulin-induced NO synthesis in platelets, decreases coagulation time and has proteolytic activity against insulin and plasmin. Serpin A1 / A1AT protects tissues from enzymes of inflammatory cells, especially neutrophil elastase. Defects in SERPINA1 are the cause of alpha-1-antitrypsin deficiency (A1ATD).
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.4 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
AAH11991 | |
50 μg | |
Glu 25 - Lys 418 | |
SerpinA1,PI,A1A,AAT,PI1,A1AT,MGC9222,PRO2275,MGC23330,alpha1AT,SPAAT | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
45.4 kDa | |
Human Serpin A1, His Tag (SE1-H5226) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Lys 418 (Accession # AAH11991). | |
-20°C | |
Serpin A1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |