Learn More
Abnova™ Human SPATA2 Partial ORF (NP_006029, 422 a.a. - 519 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque:  Abnova™ H00009825-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16151336
  • 508.00€ / 25µg
Expédition estimée: 30-05-2024
Description
Sequence: SLAHGASLREKYPGQTQGLDRLPHLHSKSKPSTTPTSRCGFCNRPGATNTCTQCSKVSCDACLSAYHYDPCYKKSELHKFMPNNQLNYKSTQLSHLVY
Spécification
Spécification
NP_006029
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SLAHGASLREKYPGQTQGLDRLPHLHSKSKPSTTPTSRCGFCNRPGATNTCTQCSKVSCDACLSAYHYDPCYKKSELHKFMPNNQLNYKSTQLSHLVY
RUO
SPATA2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9825
SPATA2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ13167/KIAA0757/PD1/tamo
SPATA2
Recombinant
wheat germ expression system