Learn More
Abnova™ Human ST6GALNAC2 Partial ORF (NP_006447, 161 a.a. - 270 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00010610-Q01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
ST6GALNAC2 belongs to a family of sialyltransferases that add sialic acids to the nonreducing ends of glycoconjugates. At the cell surface, these modifications have roles in cell-cell and cell-substrate interactions, bacterial adhesion, and protein targeting (Samyn-Petit et al., 2000 [PubMed 10742600]).[supplied by OMIM]
Sequence: NGSRQGPNIDAHDYVFRLNGAVIKGFERDVGTKTSFYGFTVNTMKNSLVSYWNLGFTSVPQGQDLQYIFIPSDIRDYVMLRSAILGVPVPEGLDKGDRPHAYFGPEASASSpécification
NP_006447 | |
Liquid | |
10610 | |
ST6GALNAC2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ45660/SAITL1/SIAT7/SIAT7B/SIATL1/ST6GalNAII/STHM | |
ST6GALNAC2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NGSRQGPNIDAHDYVFRLNGAVIKGFERDVGTKTSFYGFTVNTMKNSLVSYWNLGFTSVPQGQDLQYIFIPSDIRDYVMLRSAILGVPVPEGLDKGDRPHAYFGPEASAS | |
RUO | |
ST6GALNAC2 | |
Wheat Germ (in vitro) | |
GST |