missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ST8SIA2 Partial ORF (NP_006002, 40 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal
Code nomenclature Nacres: NA.28
Click to view available options
Quantité:
10 μg
25 μg
Conditionnement:
10µg
25µg
Description
The protein encoded by this gene is a type II membrane protein that is thought to catalyze the transfer of sialic acid from CMP-sialic acid to N-linked oligosaccharides and glycoproteins. The encoded protein may be found in the Golgi apparatus and may be involved in the production of polysialic acid, a modulator of the adhesive properties of neural cell adhesion molecule (NCAM1). This protein is a member of glycosyltransferase family 29. [provided by RefSeq]
Sequence: TIRSAVNSLHSKSNRAEVVINGSSSPAVVDRSNESIKHNIQPASSKWRHNQTLSLRIRKQILKFLDAEKDISVLKGTLKPGDIIHYIFDRDSTMNVSQNLYELLPRT
Spécification
Spécification
Numéro d’adhésion | NP_006002 |
À utiliser avec (application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Identification génétique (Entrez) | 8128 |
Poids moléculaire | 37.51kDa |
Nom | ST8SIA2 (Human) Recombinant Protein (Q01) |
Test du contrôle qualité | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantité | 25 μg |
Immunogène | TIRSAVNSLHSKSNRAEVVINGSSSPAVVDRSNESIKHNIQPASSKWRHNQTLSLRIRKQILKFLDAEKDISVLKGTLKPGDIIHYIFDRDSTMNVSQNLYELLPRT |
Conditions de stockage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Abnova™ Human ST8SIA2 Partial ORF (NP_006002, 40 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal >
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu