Learn More
Abnova™ Human SUDS3 Full-length ORF (NP_071936.2, 1 a.a. - 328 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00064426-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
SDS3 is a subunit of the histone deacetylase (see HDAC1; MIM 601241)-dependent SIN3A (MIM 607776) corepressor complex (Fleischer et al., 2003 [PubMed 12724404]).[supplied by OMIM]
Sequence: MSAAGLLAPAPAQAGAPPAPEYYPEEDEELESAEDDERSCRGRESDEDTEDASETDLAKHDEEDYVEMKEQMYQDKLASLKRQLQQLQEGTLQEYQKRMKKLDQQYKERIRNAELFLQLETEQVERNYIKEKKAAVKEFEDKKVELKENLIAELEEKKKMIENEKLTMELTGDSMEVKPIMTRKLRRRPNDPVPIPDKRRKPAPAQLNYLLTDEQIMEDLRTLNKLKSPKRPASPSSPEHLPATPAESPAQRFEARIEDGKLYYDKRWYHKSQAIYLESKDNQKLSCVISSVGANEIWVRKTSDSTKMRIYLGQLQRGLFVIRRRSAASpécification
NP_071936.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ00052/MGC104711/SAP45/SDS3 | |
SUDS3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64426 | |
SUDS3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSAAGLLAPAPAQAGAPPAPEYYPEEDEELESAEDDERSCRGRESDEDTEDASETDLAKHDEEDYVEMKEQMYQDKLASLKRQLQQLQEGTLQEYQKRMKKLDQQYKERIRNAELFLQLETEQVERNYIKEKKAAVKEFEDKKVELKENLIAELEEKKKMIENEKLTMELTGDSMEVKPIMTRKLRRRPNDPVPIPDKRRKPAPAQLNYLLTDEQIMEDLRTLNKLKSPKRPASPSSPEHLPATPAESPAQRFEARIEDGKLYYDKRWYHKSQAIYLESKDNQKLSCVISSVGANEIWVRKTSDSTKMRIYLGQLQRGLFVIRRRSAA | |
RUO | |
SUDS3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |